Anti-ITPR1 Mouse Monoclonal Antibody [clone: S24-18]

Supplier: Rockland Immunochemical
200-301-F77
RL200-301-F77EA 588.97 USD
RL200-301-F77
Anti-ITPR1 Mouse Monoclonal Antibody [clone: S24-18]
Antibodies
Anti-IP3 Antibody is suitable for use in WB, IP, and IHC. Expect a band approximately ~300kDa on specific lysates. Does not recognize type 2 or type 3 IP3R's. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 1.0-10µg/ml; Western Blot: 1µg/ml; Immunochemistry: 0.1-1.0µg/ml

Type: Primary
Antigen: ITPR1
Clonality: Monoclonal
Clone: S24-18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR