Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: Hyb-K41220A]

Supplier: Rockland Immunochemical
200-301-F75
RL200-301-F75EA 588.97 USD
RL200-301-F75
Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: Hyb-K41220A]
Antibodies
Anti-Hsp90a-beta Antibody is suitable for WB, ELISA, and IHC. Expect a band approximately 90kD Proteins corresponding to the molecular mass of hsp90 a or -beta on SDS Page immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1µg/ml

Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: Hyb-K41220A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat, S. Cerevisiae, S. Pombe
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR