Anti-YDJ1 Mouse Monoclonal Antibody [clone: 2A7.H6]

Supplier: Rockland Immunochemical
200-301-F59
RL200-301-F59EA 588.97 USD
RL200-301-F59
Anti-YDJ1 Mouse Monoclonal Antibody [clone: 2A7.H6]
Antibodies
Anti-Hsp40, YDJ1 Antibody is recommended for use in WB, IP, and ELISA. Expect a band approximately ~40kDa in specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000;

Type: Primary
Antigen: YDJ1
Clonality: Monoclonal
Clone: 2A7.H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR