Anti-HMOX1 Mouse Monoclonal Antibody [clone: 1F12-A6]

Supplier: Rockland Immunochemical
200-301-F47
RL200-301-F47EA 588.97 USD
RL200-301-F47
Anti-HMOX1 Mouse Monoclonal Antibody [clone: 1F12-A6]
Antibodies
Anti-HO-1 Antibody is suitable for WB, IP, Immunofluorescence microscopy, ELISA and IHC. Expect a band approximately 32kDa Protein, corresponding to the molecular mass of HO-1 on SDS Page immunoblots.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:500-1000;

Type: Primary
Antigen: HMOX1
Clonality: Monoclonal
Clone: 1F12-A6
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Bovine, Dog, Guinea Pig, Hamster, Monkey, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR