Anti-HCN1 Mouse Monoclonal Antibody [clone: S70-28]

Supplier: Rockland Immunochemical
200-301-F40
RL200-301-F40EA 588.97 USD
RL200-301-F40
Anti-HCN1 Mouse Monoclonal Antibody [clone: S70-28]
Antibodies
Anti-HCN1 Antibody is suitable for use in WB, IP and IHC. Expect a band approximately ~100kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions:

Type: Primary
Antigen: Hcn1
Clonality: Monoclonal
Clone: S70-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR