Anti-HSPA5 Mouse Monoclonal Antibody [clone: 6H4-2G7]

Supplier: Rockland Immunochemical
200-301-F37
RL200-301-F37EA 588.97 USD
RL200-301-F37
Anti-HSPA5 Mouse Monoclonal Antibody [clone: 6H4-2G7]
Antibodies
Type: Primary
Antigen: HSPA5
Clonality: Monoclonal
Clone: 6H4-2G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Cryphonectria Parasitica, Hamster, Monkey, Rabbit, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR