Anti-GABRD Mouse Monoclonal Antibody [clone: S151-3]

Supplier: Rockland Immunochemical
200-301-F33
RL200-301-F33EA 588.97 USD
RL200-301-F33
Anti-GABRD Mouse Monoclonal Antibody [clone: S151-3]
Antibodies
Anti-GABA-A receptor delta Antibody is suitable for use in WB and Immunofluorescence microscopy. Expect a band approximately ~55kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:5000-1:25000; Immunofluorescence: 1.0-10µg/ml; Western Blot: 1-10µg/ml; Immunochemistry: 0.1-1.0µg/ml

Type: Primary
Antigen: Gabrd
Clonality: Monoclonal
Clone: S151-3
Conjugation: Unconjugated
Epitope: Internal
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR