Anti-GABRB1 Mouse Monoclonal Antibody [clone: S96-55]

Supplier: Rockland Immunochemical
200-301-F30
RL200-301-F30EA 588.97 USD
RL200-301-F30
Anti-GABRB1 Mouse Monoclonal Antibody [clone: S96-55]
Antibodies
Anti-GABA-A receptor alpha 1 Antibody is suitable for use in WB and Immunofluorescence microscopy. Expect a band approximately ~55kDa on specific lysates. Antibody is provided in PBS pH 7.4. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 1.0-10 µg/ml; Western Blot: 1 µg/ml; Immunochemistry: 0.1-1.0 µg/ml

Type: Primary
Antigen: Gabra1
Clonality: Monoclonal
Clone: S96-55
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR