Anti-LGR4 Mouse Monoclonal Antibody [clone: 6G8.B3.G5.C3]

Supplier: Rockland Immunochemical
200-301-B45 200-301-B45S
RL200-301-B45EA 565.22 USD
RL200-301-B45 RL200-301-B45S
Anti-LGR4 Mouse Monoclonal Antibody [clone: 6G8.B3.G5.C3]
Antibodies
This monoclonal antibody is suitable for ELISA, immunochemistry and western blotting. Expect a band approximately 102 kDa in size corresponding to LGR4 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:500 - 1:3000; Immunochemistry: 5 µg/ml;

Type: Primary
Antigen: LGR4
Clonality: Monoclonal
Clone: 6G8.B3.G5.C3
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Chimpanzee, Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR