Anti-WNT1 Mouse Monoclonal Antibody [clone: 13F9]

Supplier: Rockland Immunochemical
200-301-A37S
RL200-301-A37SEA 151.49 USD
RL200-301-A37S
Anti-WNT1 Mouse Monoclonal Antibody [clone: 13F9]
Antibodies
Type: Primary
Antigen: WNT1
Clonality: Monoclonal
Clone: 13F9
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR