Anti-H5N1 Mouse Monoclonal Antibody [clone: 8A3]

Supplier: Rockland Immunochemical
200-301-978
RL200-301-978EA 565.22 USD
RL200-301-978
Anti-H5N1 Mouse Monoclonal Antibody [clone: 8A3]
Antibodies
Hemagglutinin of A/Vietnam/1203/04 Influenza Virus (VN04-10) monoclonal antibody can be used for hemagglutination inhibition (HI) assays to provide antigenic characterization of the influenza A viruses of the H5 HA subtype.

Recommended Dilutions: ELISA: 1:5000;

Type: Primary
Antigen: H5N1 (VN04-10)
Clonality: Monoclonal
Clone: 8A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: H5N1 virus, Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR