Anti-MAD2L1 Mouse Polyclonal Antibody [clone: 17D10]

Supplier: Rockland Immunochemical
200-301-461
RL200-301-461EA 565.22 USD
RL200-301-461
Anti-MAD2L1 Mouse Polyclonal Antibody [clone: 17D10]
Antibodies
This Protein A purified antibody has been tested for use in immunoprecipitation and by western blot.  This antibody is not applicable for immunofluorescence microscopy.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:200 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: MAD2L1
Clonality: Polyclonal
Clone: 17D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR