Anti-SMC1B Mouse Polyclonal Antibody [clone: 5D11.G5]

Supplier: Rockland Immunochemical
200-301-397
RL200-301-397EA 565.22 USD
RL200-301-397
Anti-SMC1B Mouse Polyclonal Antibody [clone: 5D11.G5]
Antibodies
Protein G Purified Mab anti-SMC1 was tested by ELISA,.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Immunofluorescence: 2.5 µg/ml; Western Blot: 1:100 - 1:2000;

Type: Primary
Antigen: SMC1B
Clonality: Polyclonal
Clone: 5D11.G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR