Anti-TG Mouse Polyclonal Antibody [clone: 2H11]

Supplier: Rockland Immunochemical
200-301-388
RL200-301-388EA 565.22 USD
RL200-301-388
Anti-TG Mouse Polyclonal Antibody [clone: 2H11]
Antibodies
This antibody is suitable for the detection of thyroglobulin in ELISA, immunochemistry, immunoblotting and immunoprecipitation.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:50 - 1:200; Immunochemistry: 1:50 - 1:200;

Type: Primary
Antigen: TG
Clonality: Polyclonal
Clone: 2H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR