Anti-LEFTY2 Mouse Polyclonal Antibody [clone: 7C5G1H6H10]

Supplier: Rockland Immunochemical
200-301-279
RL200-301-279EA 565.22 USD
RL200-301-279
Anti-LEFTY2 Mouse Polyclonal Antibody [clone: 7C5G1H6H10]
Antibodies
This antibody is suitable for ELISA and western blotting.  The antibody may be used for other applications, such as RIA.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:2000;

Type: Primary
Antigen: LEFTY2
Clonality: Polyclonal
Clone: 7C5G1H6H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR