Anti-HSPB1 Mouse Polyclonal Antibody [clone: G3.1]

Supplier: Rockland Immunochemical
200-301-243
RL200-301-243EA 565.22 USD
RL200-301-243
Anti-HSPB1 Mouse Polyclonal Antibody [clone: G3.1]
Antibodies
This Protein A purified monoclonal antibody against human Hsp27 has been tested for use in immunoblotting, immunoprecipitation, ELISA.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: HSPB1
Clonality: Polyclonal
Clone: G3.1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR