Anti-PFKM Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
200-1356S
RL200-1356SEA 150.22 USD
RL200-1356S
Anti-PFKM Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Fructose-6-Phosphate Kinase purified antibody has been tested for use in ELISA, immunofluorescence microscopy and western blot.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 48 kDa in size.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Do not add Sodium Azide!

Type: Primary
Antigen: PFKM
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR