Anti-GDHB Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
200-103-214S
RL200-103-214SEA 150.22 USD
RL200-103-214S
Anti-GDHB Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Glucose Dehydrogenase has been assayed against 1.0 µg of Glucose Dehydrogenase (Bacillus) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:1000 - 1:5000;
Do not add Sodium Azide!

Type: Primary
Antigen: gdhB
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bacillus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR