Anti-GOX Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
200-101-037 200-101-037S
RL200-101-037EA 551.64 USD
RL200-101-037 RL200-101-037S
Anti-GOX Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:1000 - 1:5000;

Type: Primary
Antigen: gox
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Aspergillus niger
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR