Anti-RBC Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
109-4139
RL109-4139EA 302.99 USD
RL109-4139
Anti-RBC Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-HUMAN Red Blood Cell (RBC) Reacts with Human

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml
0

Type: Primary
Antigen: RBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR