Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 14H5]

Supplier: Rockland Immunochemical
109-401-304
RL109-401-304EA 1380.15 USD
RL109-401-304
Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 14H5]
Antibodies
This antiserum against anti-Human IL-1 Receptor Type I (IL-1RI) has been tested for use in ELISA,  radioimmunoassays and immunoprecipitation.  Reactivity in other immunoassays is unknown.

Recommended Dilutions:

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: 14H5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR