Anti-RBC Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
103-4139
RL103-4139EA 298 USD
RL103-4139
Anti-RBC Rabbit Polyclonal Antibody
Antibodies
Suitable for agglutination of cells on titer plates. Each laboratory should determine an optimum working titer for use in its particular application. Other applications have not been tested but use in such assays should not necessarily be excluded.

Recommended Dilutions: Agglutination Titer 1:32 - 1:64;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: RBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR