Anti-KINESIN-1 Mouse Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-E80S
RL100401E80SEA 151.49 USD
RL100401E80S
Anti-KINESIN-1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KINESIN-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR