Anti-KTI3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-4179
RL100-4179EA 1380.15 USD
RL100-4179
Anti-KTI3 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-TRYPSIN INHIBITOR (Soy Bean) Reacts with Pan

Recommended Dilutions: ELISA: 1:3000 - 1:15000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: KTI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Soy Bean
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR