Anti-DNASE1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-4161
RL100-4161EA 1380.15 USD
RL100-4161
Anti-DNASE1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-DEOXYRIBONUCLEASE I (Bovine Pancreas) Reacts with Pan

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: DNASE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR