Anti-AAO Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-4148
RL100-4148EA 1380.15 USD
RL100-4148
Anti-AAO Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-ASCORBATE OXIDASE (cucurbita species) Reacts with Pan

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: AAO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Cucurbita
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR