Anti-ADH1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-4143
RL100-4143EA 1380.15 USD
RL100-4143
Anti-ADH1 Rabbit Polyclonal Antibody
Antibodies
This antibody has been tested for use in ELISA and by western blot.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ADH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Saccharomyces cerevisiae, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR