Anti-GLB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-4136
RL100-4136EA 329.1 USD
RL100-4136
Anti-GLB1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-BETA GALACTOSIDASE (E.coli) Reacts with Pan

Type: Primary
Antigen: Beta Galactosidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR