Anti-GM4841 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-N95S
RL100-401-N95SEA 151.49 USD
RL100-401-N95S
Anti-GM4841 Rabbit Polyclonal Antibody
Antibodies
Anti-IFN Induced GTPase Like Protein has been tested by ELISA and Western Blot. Expect a band at 48 kDa by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: GM4841
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR