Anti-ENO2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-E00
RL100-401-E00EA 578.81 USD
RL100-401-E00
Anti-ENO2 Rabbit Polyclonal Antibody
Antibodies
Anti-Neuron specific enolase (Rabbit) antibody is suitable for use in Western Blot and IF. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 47kDa in size corresponding to NSE Protein by western blotting.

Recommended Dilutions: Immunofluorescence: 1:500; Western Blot: 1:2000;

Type: Primary
Antigen: ENO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR