Anti-BIRA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-B21
RL100-401-B21EA 447.16 USD
RL100-401-B21
Anti-BIRA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BIRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR