Anti-CUL2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-A02
RL100-401-A02EA 551.64 USD
RL100-401-A02
Anti-CUL2 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human Cul2 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated product and Protein from cell lysates (using HeLa or NIH-3T3).  An 86.9 kDa band corresponding to human Cul2 is detected.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:500;

Type: Primary
Antigen: CUL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR