Anti-ZNF148 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-685
RL100-401-685EA 551.64 USD
RL100-401-685
Anti-ZNF148 Rabbit Polyclonal Antibody
Antibodies
This polyclonal antibody reacts human ZBP-89 in a variety of immunological assays including western blot and ELISA.

  • Recommended Dilutions: ELISA: 1:10000 - 1:30,000; Western Blot: 1:5000;

Type: Primary
Antigen: ZNF148
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR