Anti-NOTCH2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-408S
RL100-401-408SEA 151.49 USD
RL100-401-408S
Anti-NOTCH2 Rabbit Polyclonal Antibody
Antibodies
A strong response was detected by ELISA against the immunizing peptide.This product was assayed against the peptide immunogen in a standard capture ELISA using Peroxidase conjugated anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:400 - 1:2000; Immunochemistry: 1:500;

Type: Primary
Antigen: NOTCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR