Anti-MYC Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-224
RL100-401-224EA 551.64 USD
RL100-401-224
Anti-MYC Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human and mouse c-myc Protein, which is a nuclear Protein with a molecular weight of approximately 66 kDa.  This antiserum detects c-myc by immunoblotting assays in both overexpressed and endogenous systems.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000;

Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR