Anti-CDK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-161
RL100-401-161EA 551.64 USD
RL100-401-161
Anti-CDK2 Rabbit Polyclonal Antibody
Antibodies
Anti-cdk2 antbody is suitable for immunochemistry, western blotting and other antibody based assays.  Multiple isoforms as well as phosphorylated forms of cdk-2 may be detected by this antibody. Use paraffin embedded tissues for immunochemistry.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDK2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR