Anti-GPD1 Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
100-101-210
RL100-101-210EA 1380.15 USD
RL100-101-210
Anti-GPD1 Goat Polyclonal Antibody
Antibodies
This product has been assayed against 1.0 µg of Glycerol-3-Phosphate-Dehydrogenase (Rabbit Muscle) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: GPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR