IgG, AP (Alkaline Phosphatase)
Supplier: Rockland Immunochemical
011-0502
RL011-0502EA 228.81 USD
RL011-0502
IgG, AP (Alkaline Phosphatase)
Proteins and Peptides
Rabbit IgG Alkaline Phosphatase Conjugate
  • control or standard reagent
  • ELISA, western blot, and immunohistochemistry
Rabbit IgG alk phos conjugated whole molecule - secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-affinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present.
Rabbit IgG alk phos conjugated whole molecule suitable for ELISA, western blot, and immunohistochemistry, as well as other assays requiring lot-to-lot consistency.