Human Recombinant IL17A (from E. coli)
Supplier: Rockland Immunochemical
Interleukin17-A is a proinflammatory cytokine produced by activated T cells and exists as a homodimer. This cytokine regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-17A can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of this cytokine are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis. IL17-A is the founding member of a group of cytokines called the IL-17 family. IL17-A was originally identified as a transcript from a rodent T-cell hybridoma. To elicit its functions, IL17 binds to a type I cell surface receptor called IL17R of which there are at least three variants: IL17RA, IL17RB, and IL17RC.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)