Histatin 5
Supplier: Rockland Immunochemical
000-001-M59
RL000-001-M59EA 310.3 USD
RL000-001-M59
Histatin 5
Proteins and Peptides
Histatin Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Histatins are antimicrobial and antifungal proteins found in the saliva. They play a role in wound closure. The three major histatins are HTN1, HTN3, and HTN5.
Histatin 5 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.