HIV tat 48-60
Supplier: Rockland Immunochemical
000-001-M46
RL000-001-M46EA 124.33 USD
RL000-001-M46
HIV tat 48-60
Proteins and Peptides
HIV tat protein Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Translocation through the plasma membrane has been shown to be a major limiting step for the delivery of various macromolecules to the cytoplasm and other intracellular compartments (e.g., mitochondria, nucleus). Numerous studies have confirmed that specific peptide sequences known as cell penetrating peptides (CPP) derived from proteins able to cross the plasma membrane, can be added to various cargo and delivered across cell membranes. The cargo molecules that have been successfully transported into cells includes oligonucleotides, peptides, peptide nucleic acids, proteins and nanoparticles. One of these translocating peptides was derived from the HIV-1 Tat protein, specifically located within the first exon of the HIV tat protein. The specific HIV tat sequence is highly basic (cationic) and is readily added to peptides either as a preformed peptide with a site for direct conjugation to other molecules (typically a cysteine). Addition of the tat-cargo complex (5-50 uM concentration) to cells for 30-60 minutes results in the transfer of the tat-cargo complex to intracellular locations in a rapid, dose-dependent manner. The addition of nuclear or mitochondrial localization sequences has been shown to specifically direct the cargo to the nucleus or mitochondria respectively.
HIV-1 tat, 48-60 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.