Rat pTH 5-13
Supplier: Rockland Immunochemical
000-001-M30
RL000-001-M30EA 258.06 USD
RL000-001-M30
Rat pTH 5-13
Proteins and Peptides
Parathyroid hormone Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Parathyroid hormone (pTH) is the most important endocrine regulator of Ca2+ and phosphorus concentration in the extracellular fluid. pTH is secreted from cells of the parathyroid glands and acts for the most part via the binding to specific G-protein coupled receptors on the surface of  target cells in bone and kidney, utilizing the second messenger adenylate cyclase to regulate cAMP levels in cells.  Like most other protein hormones, parathyroid hormone is synthesized as a preprohormone and is processed intracellularly.  The mature hormone, 84 amino acids in length, is released from secretory vesicles into the blood.  Biologic effects of large C-terminal parathyroid hormone (PTH) fragments, such as pTH 7-84, are thought to regulate PTH secretion via an autocrine/paracrine regulatory mechanism
Parathyroid hormone (pTH) Rat 5-13 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.