HR23B Peptide
Supplier: Rockland Immunochemical
000-001-391
RL000-001-391EA 166.12 USD
RL000-001-391
HR23B Peptide
Proteins and Peptides
HR23B Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
HR23B peptide for use in control assays with purified antibodies Anti-HR23B (p/n 600-101-391).
Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay.