Human AP1G1 Peptide
Supplier: Rockland Immunochemical
000-001-289
RL000-001-289EA 166.12 USD
RL000-001-289
Human AP1G1 Peptide
Proteins and Peptides
Human AP1G1 Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
AP1G1 peptide for use in control assays with purified antibodies Anti-AP1G1 (p/n 600-101-289).
Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay.