AP2A Peptide
Supplier: Rockland Immunochemical
000-001-287
RL000-001-287EA 166.12 USD
RL000-001-287
AP2A Peptide
Proteins and Peptides
AP2A Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
AP2A peptide for use in control assays with purified antibodies Anti-AP2A (p/n 600-101-287).
Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay.