Anti-IgG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
609-4117
RL-609-4117EA 214.18 USD
RL-609-4117
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Antibody Anti-Human IgG (H&L) is suitable for immunoblotting (western or dot blot), ELISA, and immunohistochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:75000 - 1:500000; Western Blot: 1:10000 - 1:50000; Immunochemistry: 1:1000 - 1:10000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR