Used Motor Oil Composite Standard, Restek
Supplier: RESTEK CORP
31465
RK31465EA 45.5 USD
RK31465
Used Motor Oil Composite Standard, Restek
Standards Petroleum Industry Standards
Prepared from an equal-volume blend from five gasoline-powered vehicles. After blending, a precisely weighed amount of the composite is added to a volumetric flask to produce the standard. Motor Oil standards may exhibit lot-to-lot variation.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.