EPA method 8040 Phenol Standard No. 2, Restek
Supplier: RESTEK CORP
31089
RK31089EA 54 USD
RK31089
EPA method 8040 Phenol Standard No. 2, Restek
Standards Environmental Analysis Standards
Standard is packaged in isopropanol at a concentration of 2000 µg/mL in a 1 mL ampoule.
  • Mixture contains 2-Chlorophenol, 2,6-Dichlorophenol, 2,4-Dimethylphenol, 2,4-Dinitrophenol, sec-Butyl-4,6-dinitrophenol (Dinoseb™), 2-Methylphenol, 4-Methylphenol, 2,3,4,6-Tetrachlorophenol, and 2,4,5-Trichlorophenol
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).