Siltek® Treated Inlet Support Springs for Bruker/Varian 1075/1077 Split Injectors, Restek
Supplier: Avantor
RK21690PK 28.23 USD
RK21690
Siltek® Treated Inlet Support Springs for Bruker/Varian 1075/1077 Split Injectors, Restek
Springs
Inlet support springs are Siltek® treated to eliminate sample adsorption.
For use with Bruker/Varian 1075/1077 split injectors.