Replacement Inlet Seals for Agilent GCs, Restek
Supplier: Restek
21305 20391 21308 21316 21317 21320 20393 21307 21315 21318 21306 20390 21319 23415 23416 23417
RK20392PK 58.28 USD
RK20392 RK21305 RK20391 RK21308 RK21316 RK21317 RK21320 RK20393 RK21307 RK21315 RK21318 RK21306 RK20390 RK21319 10059-102 10856-128 10856-130
Replacement Inlet Seals for Agilent GCs, Restek
Seals Mechanical Seals
The inlet seal at the base of the Agilent 5890/6890/7890 GC injection port contacts the sample and must be changed frequently to prevent adsorption of active compounds.
  • All seals include washers
In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement
Note: The 1.2 mm inlet seal is recommended when installing two columns using a two-hole Vespel®/graphite ferrule.
To reduce breakdown and adsorption of active compounds, use Siltek® or gold-plated seals.
Accessories information: Note: The 1.2 mm inlet seal is recommended when installing two columns using a two-hole Vespel®/graphite ferrule.